Skip to Content
Merck
All Photos(1)

Key Documents

263842

Sigma-Aldrich

Triacontane

98%

Synonym(s):

n-Triacontane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)28CH3
CAS Number:
Molecular Weight:
422.81
Beilstein:
1777281
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

bp

258-259 °C/3 mmHg (lit.)

mp

64-67 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C30H62/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-30H2,1-2H3

InChI key

JXTPJDDICSTXJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triacontane is a straight-chain alkane that can be produced from palmitic acid via Kolbe electrolysis.

Interfacial molecular ordering of triacontane, at the SiO2/air interface, for submonolayer and excess coverage has been investigated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long chain n-alkanes at SiO2/air interfaces: Molecular ordering, annealing, and surface freezing of triacontane in the case of excess and submonolayer coverage.
Schollmeyer H, et al.
Langmuir, 19(12), 5042-5051 (2003)
Electrochemical conversion of palmitic acid via Kolbe electrolysis for synthesis of n-triacontane
Zhang Y, et al.
Journal of Electroanalytical Chemistry, 822, 73-80 (2018)
Scott A Harding et al.
Tree physiology, 34(11), 1240-1251 (2013-12-18)
The partitioning of carbon for growth, storage and constitutive chemical defenses is widely framed in terms of a hypothetical sink-source differential that varies with nutrient supply. According to this framework, phenolics accrual is passive and occurs in source leaves when
Rosaria Costa et al.
Journal of pharmaceutical and biomedical analysis, 117, 499-509 (2015-10-18)
Artemisia arborescens, also known as tree wormwood, is a typical species of the Mediterranean flora. It has been used in folk medicine for its antispasmodic, anti-pyretic, anti-inflammatory, and abortifacient properties. In the current study, the application of multidimensional comprehensive gas
Pavani P Nadiminti et al.
Protoplasma, 252(6), 1475-1486 (2015-02-26)
Electron microscopy techniques such as transmission electron microscopy (TEM) and scanning electron microscopy (SEM) have been invaluable tools for the study of the micromorphology of plant cuticles. However, for electron microscopy, the preparation techniques required may invariably introduce artefacts in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service