Skip to Content
Merck
All Photos(3)

Key Documents

327190

Sigma-Aldrich

Isobutyl nitrite

95%

Synonym(s):

2-Methylpropyl nitrite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2ONO
CAS Number:
Molecular Weight:
103.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.373 (lit.)

bp

66-67 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

functional group

nitroso

storage temp.

2-8°C

SMILES string

CC(C)CON=O

InChI

1S/C4H9NO2/c1-4(2)3-7-5-6/h4H,3H2,1-2H3

InChI key

APNSGVMLAYLYCT-UHFFFAOYSA-N

General description

Isobutyl nitrite was extensively used as room odorizers. It is a popular recreational drug.

Application

Isobutyl nitrite was used to study inhalant-induced immunotoxicity.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Flam. Liq. 2 - Muta. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-5.8 °F - closed cup

Flash Point(C)

-21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Kielbasa et al.
AAPS pharmSci, 2(2), E11-E11 (2001-12-14)
Our objective was to examine the pharmacokinetic/hemodynamic properties of inhaled isobutyl nitrite (ISBN) in rats. ISBN is one of the volatile organic nitrites that has been used primarily as a drug of abuse. Recent studies indicate, however, that these compounds
Inhaled isobutyl nitrite produced lung inflammation with increased macrophage TNF-alpha and nitric oxide production.
L S Soderberg et al.
Advances in experimental medicine and biology, 402, 187-189 (1996-01-01)
Review of the physiological effects of amyl, butyl, and isobutyl nitrites.
T J Haley
Clinical toxicology, 16(3), 317-329 (1980-05-01)
L S F Soderberg et al.
Toxicology letters, 152(1), 47-56 (2004-08-06)
Epidemiological studies have identified abuse of nitrite inhalants as an independent co-factor in HIV infection and in Kaposi's sarcoma (KS) in AIDS patients. In the present study we investigated the ability of macrophages from mice exposed to isobutyl nitrite to
L S Soderberg et al.
Experimental hematology, 24(7), 848-853 (1996-06-01)
Isobutyl nitrite is representative of a group of inhalants abused primarily by male homosexuals; abuse of this drug may be a risk factor for AIDS or Kaposi's sarcoma. Using a 14-day exposure regimen, we previously reported that inhaled isobutyl nitrite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service