Skip to Content
Merck
All Photos(2)

Key Documents

226637

Sigma-Aldrich

Butyl nitrite

95%

Synonym(s):

n-Butyl nitrite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3ONO
CAS Number:
Molecular Weight:
103.12
Beilstein:
1701036
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

760 mmHg ( 78 °C)

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.376 (lit.)

bp

78 °C (lit.)

solubility

alcohol: miscible(lit.)
diethyl ether: miscible(lit.)

density

0.882 g/mL at 25 °C (lit.)

functional group

O-nitroso
nitroso

storage temp.

2-8°C

SMILES string

CCCCON=O

InChI

1S/C4H9NO2/c1-2-3-4-7-5-6/h2-4H2,1H3

InChI key

JQJPBYFTQAANLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The photodissociation dynamics of butyl nitrite was studied using time-resolved Fourier transform infrared (TR-FTIR) emission spectroscopy. The effects of butyl nitrite on methyl cobalamin and 5-methyl tetrahydrofolate were studied.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

8.6 °F - closed cup

Flash Point(C)

-13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L D Fechter et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 12(1), 56-61 (1989-01-01)
Butyl nitrite is the predominant and presumed active ingredient in a variety of commercial preparations sold as "room odorizers." These compounds have significant abuse potential, giving the user the sensation of a "rush", which may be related to their intense
M Abu Khaled et al.
Biochemical and biophysical research communications, 135(1), 201-207 (1986-02-26)
The effects of butyl nitrite, a frequently used recreational drug, on methyl cobalamin and 5-methyl tetrahydrofolate were investigated by using 1H and 31P NMR spectroscopies. While no effect could be observed in organic solvents, strong interactions of butyl nitrite with
D P McFadden et al.
Journal of applied toxicology : JAT, 5(3), 134-139 (1985-06-01)
Mice were exposed by inhalation to n-butyl, iso-butyl sec-butyl or tert-butyl nitrite in a dynamic airflow chamber 7 h per day for 60 days at concentrations that caused less than 20% fatalities. Under these conditions, body-weight gain was depressed over
Health hazards of nitrite inhalants.
H W Haverkos et al.
The American journal of medicine, 84(3 Pt 1), 479-482 (1988-03-01)
Chi-Ming Ho et al.
Chemical communications (Cambridge, England), 47(38), 10776-10778 (2011-09-03)
Nitric oxide-releasing ruthenium nanoparticles were synthesized by the reaction of alkanethiolate-protected ruthenium nanoparticles with tert-butyl nitrite ((t)BuONO), and their water-soluble derivatives are able to deliver NO to proteins such as reduced myoglobin upon light irradiation in aqueous media.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service