T72001
Trimethylacetonitrile
98%
Synonym(s):
Pivalonitrile, tert-Butyl cyanide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)3CCN
CAS Number:
Molecular Weight:
83.13
Beilstein:
1361449
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
refractive index
n20/D 1.377 (lit.)
bp
105-106 °C (lit.)
mp
15-16 °C (lit.)
density
0.752 g/mL at 25 °C (lit.)
SMILES string
CC(C)(C)C#N
InChI
1S/C5H9N/c1-5(2,3)4-6/h1-3H3
InChI key
JAMNHZBIQDNHMM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
39.2 °F - closed cup
Flash Point(C)
4 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Karolina A Tarach et al.
Molecules (Basel, Switzerland), 25(12) (2020-06-26)
An adequately tuned acid wash of hierarchical ZSM-5 zeolites offers a levelling up in the catalytic cracking of low-density polyethylene. Identification of crucial and limiting factors governing the activity of the zeolite was extended with studies about the accessibility of
Kevin Colizza et al.
Journal of the American Society for Mass Spectrometry, 27(11), 1796-1804 (2016-09-01)
Efforts to analyze trace levels of cyclic peroxides by liquid chromatography/mass spectrometry gave evidence that acetonitrile suppressed ion formation. Further investigations extended this discovery to ketones, linear peroxides, esters, and possibly many other types of compounds, including triazole and menadione.
Andrey V Markov et al.
International journal of molecular sciences, 21(10) (2020-05-21)
A series of novel 18βH-glycyrrhetinic acid (GA) derivatives containing 3'-(alkyl/phenyl/pyridin(-2″, -3″, and -4″)-yl)-1',2',4'-oxadiazole moieties at the C-30 position were synthesized by condensation of triterpenoid's carboxyl group with corresponding amidoximes and further cyclization. Screening of the cytotoxicity of novel GA derivatives
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service