Skip to Content
Merck
All Photos(1)

Key Documents

325481

Sigma-Aldrich

Furfuryl glycidyl ether

96%

Synonym(s):

2-((2,3-Epoxypropoxy)methyl)furan, furfurylglycidyl ether, 2,3-Epoxypropyl 2-furylmethyl ether, 2-[(Oxiranylmethoxy)methyl]furan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O3
CAS Number:
Molecular Weight:
154.16
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

liquid

refractive index

n20/D 1.481 (lit.)

bp

103-104 °C/11 mmHg (lit.)

density

1.122 g/mL at 25 °C

SMILES string

C(OCc1ccco1)C2CO2

InChI

1S/C8H10O3/c1-2-7(10-3-1)4-9-5-8-6-11-8/h1-3,8H,4-6H2

InChI key

RUGWIVARLJMKDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • The synthesis of multifunctional cellulose graft alternating copolymers of 3,4-dihydrocoumarin and epoxides in DBU/DMSO/CO(2) solvent system.: This study outlines the development of cellulose-based graft copolymers for potential use in advanced material applications, highlighting the incorporation of furfuryl glycidyl ether for improved performance (Guo et al., 2023).
  • High-Performance Reversible Furan-Maleimide Resins Based on Furfuryl Glycidyl Ether and Bismaleimides.: Research on novel resin systems employing furfuryl glycidyl ether to enhance reversibility and performance in composite materials, with applications in aerospace and automotive industries (Wang et al., 2023).
  • Multi-Armed Star-Shaped Block Copolymers of Poly(ethylene glycol)-Poly(furfuryl glycidol) as Long Circulating Nanocarriers.: This publication discusses the use of furfuryl glycidyl ether in the creation of block copolymers for drug delivery systems, emphasizing the enhancement of circulation times and biocompatibility (Nakagawa et al., 2023).
  • Structure and Properties of Epoxy Polysulfone Systems Modified with an Active Diluent.: Investigation into the effects of furfuryl glycidyl ether as a modifier in epoxy systems, aiming to improve the mechanical properties and heat resistance of the resulting polymers, suitable for industrial applications (Petrova et al., 2022).
  • Star-Shaped Poly(furfuryl glycidyl ether)-Block-Poly(glyceryl glycerol ether) as an Efficient Agent for the Enhancement of Nifuratel Solubility and for the Formation of Injectable and Self-Healable Hydrogel Platforms for the Gynaecological Therapies.: Details a unique use of furfuryl glycidyl ether in medical hydrogel platforms, offering potential advancements in gynecological treatments and drug delivery systems (Ziemczonek et al., 2021).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Bi et al.
Carbohydrate polymers, 212, 368-377 (2019-03-06)
Injectable thermosensitive hydrogels crosslinked physically have been extensively studied as scaffolds in biomedical field, however, their low gel stability with weak strength limits their potential applications. Here, a novel thermosensitive furyl-modified hydroxypropyl chitin polymer was synthesized homogeneously in aqueous solution
Shweta AnilKumar et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 107(2), 314-323 (2018-04-16)
Three-dimensional bioprinting is an innovative technique in tissue engineering, to create layer-by-layer structures, required for mimicking body tissues. However, synthetic bioinks do not generally possess high printability and biocompatibility at the same time. So, there is an urgent need for
Beata Strachota et al.
Polymers, 11(6) (2019-05-31)
Reversible Diels-Alder (DA) type networks were prepared from furan and maleimide monomers of different structure and functionality. The factors controlling the dynamic network formation and their properties were discussed. Evolution of structure during both dynamic nonequilibrium and isothermal equilibrium network

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service