160458
N,N′-(1,3-Phenylene)dimaleimide
97%
Synonym(s):
1,1′-(1,3-Phenylene)bis[1H -pyrrole-2,5-dione], 1,3-Bismaleimidobenzene, 1,3-Phenylenebismaleimide, 1-[3-(2,5-Dioxopyrrol-1-yl)phenyl]pyrrole-2,5-dione, N ,N ′-1,3-Phenylenebismaleimide, N ,N -m -Phenylene dimaleimide, m -Phenylenebismaleimide
About This Item
Recommended Products
Quality Level
Assay
97%
mp
198-201 °C (lit.)
SMILES string
O=C1C=CC(=O)N1c2cccc(c2)N3C(=O)C=CC3=O
InChI
1S/C14H8N2O4/c17-11-4-5-12(18)15(11)9-2-1-3-10(8-9)16-13(19)6-7-14(16)20/h1-8H
InChI key
IPJGAEWUPXWFPL-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 4 Oral
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service