Skip to Content
Merck
All Photos(2)

Key Documents

317845

Sigma-Aldrich

Nonafluoro-1-iodobutane

98%

Synonym(s):

1-Iodoperfluorobutane, Perfluorobutyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)3I
CAS Number:
Molecular Weight:
345.93
Beilstein:
1777546
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.3285 (lit.)

density

2.01 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)I

InChI

1S/C4F9I/c5-1(6,3(9,10)11)2(7,8)4(12,13)14

InChI key

PGRFXXCKHGIFSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario Bracker et al.
Molecules (Basel, Switzerland), 25(7) (2020-04-05)
In an effort to explain the experimentally observed variation of the photocatalytic activity of t Bu 3 P, n Bu 3 P and (MeO) 3 P in the blue-light regime [Helmecke et al., Org. Lett. 21 (2019) 7823], we have
Ilona Bereczki et al.
ChemMedChem, 15(17), 1661-1671 (2020-07-12)
The limited scope of antiviral drugs and increasing problem of antiviral drug resistance represent a global health threat. Glycopeptide antibiotics and their lipophilic derivatives have emerged as relevant inhibitors of diverse viruses. Herein, we describe a new strategy for the
Chang Wang et al.
Environmental health perspectives, 120(1), 119-125 (2011-10-13)
Polyfluorinated iodine alkanes (PFIs) are important intermediates in the synthesis of organic fluoride products. Recently, PFIs have been detected in fluoropolymers as residual raw materials, as well as in the ambient environment. High production volumes and potential environmental releases of
Daniel Rauber et al.
Physical chemistry chemical physics : PCCP, 19(40), 27251-27258 (2017-10-11)
Ionic liquids (ILs) exhibit tunable behaviour and properties that are due to their supramolecular structure. We synthesized a series of alkylated and fluorinated phosphonium dicyanamide ILs to study the relation between molecular structure and assembly with a focus on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service