Skip to Content
Merck
All Photos(1)

Key Documents

370525

Sigma-Aldrich

1-Iodo-1H,1H,2H,2H-perfluorodecane

96%

Synonym(s):

1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane, 1-Iodo-1H,1H,2H,2H-perfluorodecane, 1H,1H,2H,2H-Perfluoro-1-decyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)7CH2CH2I
CAS Number:
Molecular Weight:
574.02
Beilstein:
1894667
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

mp

54-58 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCI

InChI

1S/C10H4F17I/c11-3(12,1-2-28)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)9(23,24)10(25,26)27/h1-2H2

InChI key

XVKJSLBVVRCOIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Iodo-1H,1H,2H,2H-perfluorodecane was identified as Persistent Organic Pollutant (POP) and was listed as high-production-volume chemical (HPVC).

Application

1-Iodo-1H,1H,2H,2H-perfluorodecane may be used in the preparation of 2,2′-bipyridines featuring fluorinated alkyl groups.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorous modification of 2, 2'-bipyridine.
Bennett BL, et al.
Journal of Heterocyclic Chemistry, 127(1), 140-145 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service