Skip to Content
Merck
All Photos(1)

Key Documents

171441

Sigma-Aldrich

Trifluoroiodomethane

99%

Synonym(s):

Iodotrifluoromethane, Perfluoromethyl iodide, Trifluoromethyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CF3I
CAS Number:
Molecular Weight:
195.91
Beilstein:
1732740
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

bp

−22.5 °C (lit.)

mp

<−78 °C (lit.)

SMILES string

FC(F)(F)I

InChI

1S/CF3I/c2-1(3,4)5

InChI key

VPAYJEUHKVESSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Reagent used in the rhodium-catalyzed α-trifluoromethylation of α,ß-unsaturated ketones.

Packaging

Supplied in a Sure/Pac cylinder and has a brass needle valve with a male 1/4" NPTF outlet thread installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - Ozone 1 - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

D E Dodd et al.
Inhalation toxicology, 11(11), 1041-1055 (1999-11-24)
CF(3)I is being considered by the U.S. Air Force as a replacement for Halon 1301 for fire-extinguishing requirements in unoccupied spaces. The purpose of this study was to determine and evaluate the potential for CF(3)I to produce reproductive toxicity and
C Stefan Lehmann et al.
Faraday discussions, 153, 173-187 (2011-01-01)
The control of multichannel ionic fragmentation dynamics in CF3I is studied by femtosecond pulse shaping and velocity map photoelectron photoion coincidence imaging. When CF3I is photoexcited with femtosecond laser pulses around 540 nm there are two major ions observed in
D E Dodd et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 35(1), 64-77 (1997-01-01)
Trifluoroiodomethane (CF3I) is being considered as a replacement compound for halon fire suppressants. Its structure is similar to that of Halon 1301 (CF3Br), but it has very low ozone depletion potential compared to CF3Br. As part of the process of
Discovering and verifying elusive fullerene cage isomers: structures of C2-p11-(C74-D3h)(CF3)12 and C2-p11-(C(78)-D3h5)(CF3)12.
Natalia B Shustova et al.
Angewandte Chemie (International ed. in English), 46(22), 4111-4114 (2007-04-25)
Kelvin Anggara et al.
Science advances, 4(10), eaau2821-eaau2821 (2018-10-13)
Collision geometry is central to reaction dynamics. An important variable in collision geometry is the miss-distance between molecules, known as the "impact parameter." This is averaged in gas-phase molecular beam studies. By aligning molecules on a surface prior to electron-induced

Articles

In 2001, Professor William Dolbier, Jr., at the University of Florida reported1 an approach to nucleophilic trifluoromethylation based on the generation of a trifluoromethyl anion using CF3I in the presence of a powerful twoelectron reductant, tetrakis(dimethylamino)ethylene (TDAE)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service