Skip to Content
Merck
All Photos(1)

Key Documents

317276

Sigma-Aldrich

Chloroacetaldehyde solution

~55 wt. % in H2O

Synonym(s):

α-Chloroacetaldehyde, 2-Chloro-1-ethanal, 2-Chloroacetaldehyde, 2-Chloroethanal, Monochloroacetaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CHO
CAS Number:
Molecular Weight:
78.50
Beilstein:
1071226
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

~55 wt. % in H2O

refractive index

n20/D 1.4036

density

1.236 g/mL at 25 °C

functional group

aldehyde

SMILES string

[H]C(=O)CCl

InChI

1S/C2H3ClO/c3-1-2-4/h2H,1H2

InChI key

QSKPIOLLBIHNAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chloroacetaldehyde solution can be used for the synthesis of:
  • Etheno derivatives of nucleotides.
  • 2,3-Disubstituted pyrroles by Hantzsch reaction.
  • Aminotetrahydronaphthalene ketopiperazine scaffolds as potent MCH-R1 antagonists.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of leukocyte DNA 6-thioguanine nucleotide levels by high-performance liquid chromatography with fluorescence detection
Olesen KM, et al.
Journal of Chromatography. B, Biomedical Applications, 864(1-2), 149-155 (2008)
Novel synthesis of [13C4, 15N] 1H-pyrrole-2, 3, 5-tricarboxylic acid: an important biomarker for melatonin metabolism
Skaddan MB
Journal of Labelled Compounds & Radiopharmaceuticals, 53(2), 73-77 (2010)
The efficacy and cardiac evaluation of aminomethyl tetrahydronaphthalene ketopiperazines: a novel class of potent MCH-R1 antagonists
Mendez-Andino JL, et al.
Bioorganic & Medicinal Chemistry, 15(5), 2092-2105 (2007)
Stephanie L MacAllister et al.
Chemico-biological interactions, 202(1-3), 259-266 (2012-12-12)
Cyclophosphamide and ifosfamide are two commonly used DNA-alkylating agents in cancer chemotherapy that undergo biotransformation to several toxic and non-toxic metabolites, including acrolein and chloroacetaldehyde (CAA). Acrolein and CAA toxicities occur by several different mechanisms, including ROS formation and protein
Min Young Kim et al.
Chemical research in toxicology, 20(8), 1075-1083 (2007-07-31)
2-Chloroacetaldehyde (CAA), a metabolite of the carcinogen vinyl chloride, reacts with DNA to form cyclic etheno ()-lesions. AlkB, an iron-/alpha-ketoglutarate-dependent dioxygenase, repairs 1, N (6)-ethenodeoxyadenosine (A) and 3, N (4)-ethenodeoxycytidine (C) in site-specifically modified single-stranded viral genomes in vivo and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service