Skip to Content
Merck
All Photos(2)

Key Documents

185744

Sigma-Aldrich

2-Chloroethanol

99%

Synonym(s):

Ethylene chlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2OH
CAS Number:
Molecular Weight:
80.51
Beilstein:
878139
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.78 (vs air)

Quality Level

vapor pressure

5 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

797 °F

expl. lim.

16 %

refractive index

n20/D 1.441 (lit.)

bp

129 °C (lit.)

mp

−89 °C (lit.)

solubility

alcohol: miscible
water: miscible

density

1.201 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

OCCCl

InChI

1S/C2H5ClO/c3-1-2-4/h4H,1-2H2

InChI key

SZIFAVKTNFCBPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloroethanol is a versatile intermediate in the synthesis of pharmaceuticals, agrochemicals, and specialty chemicals. Photodissociation of 2-chloroethanol in a molecular beam at 193nm has been investigated. It is widely used as an industrial solvent.

Application

2-Chloroethanol was used in the synthesis of the polyethylene terepthalate model substrate, bis(benzoyloxyethyl)terephthalate.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photodissociation of 2-bromoethanol and 2-chloroethanol at 193 nm.
Hintsa EJ, et al.
J. Chem. Phys. , 92(4), 2280-2286 (1990)
Stereoregular poly(methyl methacrylate) with double-clickable ?-end: synthesis and click reaction
Y Kohsaka, et al.
Polymer Chemistry: Introduction to an Indispensable Science, 6, 3601-3607 (2015)
Bacterial degradation of 2-chloroethanol proceeds via 2-chloroacetic acid.
Stucki G and Leisinger T.
FEMS Microbiology Letters, 16(1), 123-126 (1983)
New enzymes with potential for PET surface modification.
Fischer-Colbrie G, et al.
Biocatalysis and Biotransformation, 22(5-6), 341-346 (2004)
Ernest Marco-Urrea et al.
Chemosphere, 70(3), 404-410 (2007-08-19)
The white-rot fungus Trametes versicolor degraded trichloroethylene (TCE), a highly oxidized chloroethene, and produced 2,2,2-trichloroethanol and carbon dioxide as the main products of degradation, based on the results obtained using [13C]-TCE as the substrate. For a range of concentrations of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service