Skip to Content
Merck
All Photos(1)

Key Documents

275581

Sigma-Aldrich

6-Aminoquinoline

98%

Synonym(s):

6-Quinolinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2
CAS Number:
Molecular Weight:
144.17
Beilstein:
113320
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

146 °C/0.3 mmHg (lit.)

mp

115-119 °C (lit.)

SMILES string

Nc1ccc2ncccc2c1

InChI

1S/C9H8N2/c10-8-3-4-9-7(6-8)2-1-5-11-9/h1-6H,10H2

InChI key

RJSRSRITMWVIQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Aminoquinoline was used as an internal standard in determining serum nicotine and cotinine simultaneously by using high-performance liquid chromatography (HPLC)-fluorometric detection with a postcolumn ultraviolet-photoirradiation system. It was also used as a fluorescent derivatizing agent for the detection of biochemicals and in the synthesis of tertiary N-methylated enaminonesa.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jong Bae Seo et al.
PloS one, 10(12), e0144432-e0144432 (2015-12-15)
Hydrolysis of phosphatidylinositol 4,5-bisphosphate (PIP2) of the plasma membrane by phospholipase C (PLC) generates two critical second messengers, inositol-1,4,5-trisphosphate and diacylglycerol. For the enzymatic reaction, PIP2 binds to positively charged amino acids in the pleckstrin homology domain of PLC. Here
Pierre-Jean Aragon et al.
Chemical & pharmaceutical bulletin, 52(6), 659-663 (2004-06-10)
Indoloquinoline alkaloid cryptolepine and pyridocarbazole alkaloid ellipticine are of great interest because in vitro and in vivo studies revealed their good cytotoxic properties. In order to obtain some biologically active analogs of these compounds, we developped a synthesis based on
Bryan P Ruddy et al.
Journal of controlled release : official journal of the Controlled Release Society, 306, 83-88 (2019-06-01)
Subcutaneous delivery of nicotine was performed using a novel electrically-operated needle-free jet injector, and compared to hypodermic needle delivery in a porcine model. Nicotine was delivered as a single, one-milligram dose into the abdominal skin, formulated as a 50 microliter
W Nashabeh et al.
Journal of chromatography, 600(2), 279-287 (1992-05-29)
The electrophoretic behavior of derivatized linear and branched oligosaccharides from various sources was examined in capillary zone electrophoresis with polyether-coated fused-silica capillaries. Two UV-absorbing (also fluorescent) derivatizing agents (2-aminopyridine and 6-aminoquinoline) were utilized for the electrophoresis and sensitive detection of
S A Cohen et al.
Analytical biochemistry, 211(2), 279-287 (1993-06-01)
A highly reactive amine derivatizing reagent, 6-aminoquinolyl-N-hydroxysuccinimidyl carbamate, has been synthesized. In a rapid, one-step procedure, the compound reacts with amino acids to form stable unsymmetric urea derivatives which are readily amenable to analysis by reversed phase HPLC. Studies on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service