All Photos(1)
About This Item
Linear Formula:
CH3CH2CH2SO2Cl
CAS Number:
Molecular Weight:
142.60
Beilstein:
1747500
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
4.9 (vs air)
Assay
97%
form
liquid
refractive index
n20/D 1.453 (lit.)
bp
78-79 °C/15 mmHg (lit.)
density
1.267 g/mL at 25 °C (lit.)
SMILES string
CCCS(Cl)(=O)=O
InChI
1S/C3H7ClO2S/c1-2-3-7(4,5)6/h2-3H2,1H3
InChI key
KPBSJEBFALFJTO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
1-Propanesulfonyl chloride was used in synthesis of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
206.6 °F - closed cup
Flash Point(C)
97 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Marijana Blesic et al.
Physical chemistry chemical physics : PCCP, 11(39), 8939-8948 (2010-05-08)
A set of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids, [C(n)mim][C(k)SO(3)], formed by the variation of the alkyl chain lengths both in the cation and the anion (n = 1-6, 8, or 10; k = 1-4, or 6), was synthesised, with sixteen of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service