Skip to Content
Merck
All Photos(2)

Key Documents

664766

Sigma-Aldrich

4-Methylbenzylsulfonyl chloride

96%

Synonym(s):

(4-Methylphenyl)methanesulfonyl chloride, 4-Methylbenzenemethanesulfonyl chloride, p-Methyl-α-toluenesulfonyl chloride, p-Tolylmethanesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3CC6H4CH2SO2Cl
CAS Number:
Molecular Weight:
204.67
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

96%

form

solid

mp

76-80 °C

storage temp.

2-8°C

SMILES string

Cc1ccc(CS(Cl)(=O)=O)cc1

InChI

1S/C8H9ClO2S/c1-7-2-4-8(5-3-7)6-12(9,10)11/h2-5H,6H2,1H3

InChI key

JALKUHLLMWYIAT-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service