Skip to Content
Merck
All Photos(1)

Key Documents

171506

Sigma-Aldrich

4-Hydroxybenzenesulfonic acid solution

65 wt. % in H2O

Synonym(s):

Phenol-4-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4SO3H
CAS Number:
Molecular Weight:
174.17
Beilstein:
1869034
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

concentration

65 wt. % in H2O

refractive index

n20/D 1.489

density

1.337 g/mL at 25 °C

functional group

sulfonic acid

SMILES string

Oc1ccc(cc1)S(O)(=O)=O

InChI

1S/C6H6O4S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4,7H,(H,8,9,10)

InChI key

FEPBITJSIHRMRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Lambeir et al.
European journal of biochemistry, 163(1), 123-127 (1987-02-16)
For the first time elementary reactions involving chloroperoxidase compounds I and II have been investigated. A multi-mixing stopped-flow apparatus was used to study the kinetics of the reactions of compounds I and II with ascorbic acid, ferrocyanide and p-phenolsulfonic acid.
Makoto Hagiwara et al.
Mutation research, 603(2), 111-120 (2006-01-13)
To assess the genotoxicity of 14 chemical agents used as locally applied agents in dental practice, the ability of these agents to elicit chromosome aberrations was examined using Syrian hamster embryo (SHE) cells. Chromosome aberrations in SHE cells were induced
[Effect of phenolsulfonic acid on cultured V79 cells].
T Yokka
Shigaku = Odontology; journal of Nihon Dental College, 75(1), 50-62 (1987-06-01)
Operational stability of immobilised horseradish peroxidase in mini-packed bed bioreactors.
Journal of Molecular Catalysis. B, Enzymatic, 28(2), 121-128 (2004)
Toxicity and uptake potential of an acrylic polymer comonomer and its biological degradation products.
F R Johannsen et al.
Bulletin of environmental contamination and toxicology, 40(3), 325-331 (1988-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service