Skip to Content
Merck
All Photos(1)

Key Documents

282510

Sigma-Aldrich

2-Methyl-1-phenyl-1-propene

99%

Synonym(s):

(2-Methylpropenyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=C(CH3)2
CAS Number:
Molecular Weight:
132.20
Beilstein:
1902046
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.539 (lit.)

bp

187-188 °C (lit.)

mp

−50-−48 °C (lit.)

density

0.901 g/mL at 25 °C (lit.)

functional group

phenyl

storage temp.

2-8°C

SMILES string

C\C(C)=C/c1ccccc1

InChI

1S/C10H12/c1-9(2)8-10-6-4-3-5-7-10/h3-8H,1-2H3

InChI key

BTOVVHWKPVSLBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methyl-1-phenyl-1-propene has been used in the preparation of 2-bromo-2-methyl-1-phenylpropan-1-ol and 1-bromo-2-methyl-1-phenylpropan-2-ol.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bromohydrin formation in dimethyl sulfoxide.
Dalton DR, et al.
Journal of the American Chemical Society, 90(20), 5498-5501 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service