282073
3-Amino-1,2,4-triazole-5-carboxylic acid hydrate
98%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H4N4O2 · xH2O
CAS Number:
Molecular Weight:
128.09 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
solid
mp
182 °C (lit.)
functional group
carboxylic acid
SMILES string
[H]O[H].Nc1n[nH]c(n1)C(O)=O
InChI
1S/C3H4N4O2.H2O/c4-3-5-1(2(8)9)6-7-3;/h(H,8,9)(H3,4,5,6,7);1H2
InChI key
AGJCUQSNRFXRDU-UHFFFAOYSA-N
Application
3-Amino-1,2,4-triazole-5-carboxylic acid (AmTAZAc) has been used in the synthesis of new three-dimensional metal-organic framework, [ZnF(AmTAZ)] solvents.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cheng-Yong Su et al.
Journal of the American Chemical Society, 126(11), 3576-3586 (2004-03-18)
An effective solvothermal procedure has been developed to synthesize the new three-dimensional metal-organic framework, [ZnF(AmTAZ)].solvents, using either 3-amino-1,2,4-triazole (AmTAZ) or 3-amino-1,2,4-triazole-5-carboxylic acid (AmTAZAc) and a choice of several Zn(II) salts as starting materials. The three-dimensional structure displays open-ended, hollow nanotubular
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service