Skip to Content
Merck
All Photos(1)

Key Documents

260150

Sigma-Aldrich

Glyoxylic acid solution

50 wt. % in H2O

Synonym(s):

Formylformic acid, Oxoethanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHCCOOH
CAS Number:
Molecular Weight:
74.04
Beilstein:
741891
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

concentration

50 wt. % in H2O

refractive index

n20/D 1.4149

density

1.342 g/mL at 25 °C

functional group

aldehyde

SMILES string

OC(=O)C=O

InChI

1S/C2H2O3/c3-1-2(4)5/h1H,(H,4,5)

InChI key

HHLFWLYXYJOTON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glyoxylic acid has been employed:
  • as reducing agent in electroless copper depositions by free-formaldehyde method
  • in synthesis of new chelating agent, 2-(2-((2-hydroxybenzyl)amino)ethylamino)-2-(2-hydroxyphenyl)acetic acid (DCHA)

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra López-Rayo et al.
Journal of agricultural and food chemistry, 58(13), 7908-7914 (2010-06-10)
Iron chelates analogous to o,o-EDDHA/Fe(3+) are the fertilizers chosen to treat iron chlorosis in plants growing on calcareous soil. The isomer o,p-EDDHA/Fe(3+) presents less stability but faster assimilation by the plant than o,o-EDDHA/Fe(3+), because only five coordinating groups are able
Sang-Jin Cho et al.
Journal of nanoscience and nanotechnology, 11(6), 5328-5333 (2011-07-21)
Microwave (MW) plasma was applied to the surface of polyimide (PI) films as a treatment to enhance the adhesion between copper deposition layer and PI surface for electroless plating. The influences of nitrogen MW plasma treatment on chemical composition of
En-Zhi Shen et al.
Nature, 508(7494), 128-132 (2014-02-14)
It has been theorized for decades that mitochondria act as the biological clock of ageing, but the evidence is incomplete. Here we show a strong coupling between mitochondrial function and ageing by in vivo visualization of the mitochondrial flash (mitoflash)
Ruth Belostotsky et al.
Journal of molecular medicine (Berlin, Germany), 90(12), 1497-1504 (2012-06-26)
Perturbations in glyoxylate metabolism lead to the accumulation of oxalate and give rise to primary hyperoxalurias, recessive disorders characterized by kidney stone disease. Loss-of-function mutations in HOGA1 (formerly DHDPSL) are responsible for primary hyperoxaluria type III. HOGA1 is a mitochondrial
A Arvesen et al.
VASA. Zeitschrift fur Gefasskrankheiten, 28(2), 85-94 (1999-07-20)
These experiments aimed to study the in vivo short and long term neurovascular regeneration after frostbite. The rabbit central ear-artery was used as the experimental model. The effects on the noradrenergic innervation of the artery were measured in isolated vascular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service