8.14084
Glyoxylic acid
(50% solution in water) for synthesis
Synonym(s):
Glyoxalic acid
About This Item
vapor pressure
10 hPa ( 25 °C)
Quality Level
form
liquid
potency
2528 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)
pH
0.3 (19.5 °C in H2O)
bp
111 °C/1013 hPa
mp
-93 °C
density
1.34 g/cm3 at 20 °C
storage temp.
2-30°C
Application
- Advancing mesoporous carbon synthesis for supercapacitors: Research highlights the use of glyoxylic acid as a cross-linking agent in the synthesis of mesoporous carbons, enhancing their functionality for supercapacitor applications. This study provides key insights into the design of advanced energy storage materials (Song et al., 2023).
- Thermoset films and coatings: Glyoxylic acid is utilized to create ductile, high-lignin-content thermoset films and coatings, showcasing its applicability in sustainable material engineering. This approach offers new opportunities for the development of environmentally friendly coatings and films (Boarino et al., 2023).
- Organic synthesis: A novel decarboxylative N-formylation of amines using glyoxylic acid, promoted by H2O2, has been documented, highlighting its role as a reagent in organic synthesis. This method represents a significant advancement in the synthesis of amine derivatives (Wu et al., 2023).
- Enhancing luminescence in covalent organic frameworks: A study demonstrates the incorporation of glyoxylic acid as a linker in two-dimensional covalent organic frameworks, significantly boosting their luminescence. This finding opens up new avenues for the use of these materials in optoelectronic applications (Yang et al., 2023).
Analysis Note
Density (d 20 °C/ 4 °C): 1.330 - 1.340
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - Skin Sens. 1
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service