Skip to Content
Merck
All Photos(1)

Key Documents

253715

Sigma-Aldrich

p-Tolyl isothiocyanate

97%

Synonym(s):

4-Methylphenyl isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NCS
CAS Number:
Molecular Weight:
149.21
Beilstein:
386032
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

refractive index

n20/D 1.6345 (lit.)

bp

237 °C (lit.)
62 °C/0.3 mmHg (lit.)

mp

25-26 °C (lit.)

functional group

isothiocyanate

SMILES string

Cc1ccc(cc1)N=C=S

InChI

1S/C8H7NS/c1-7-2-4-8(5-3-7)9-6-10/h2-5H,1H3

InChI key

ABQKHKWXTUVKGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Tolyl isothiocyanate reacts with 6-hydroxy-2,4,5-triaminopyrimidine or with 4,5,6-triaminopyrimidine in DMF and triethylamine to yield 2,4-diamino-5-(p-tolylthioureido)aminopyrimidin-6-one and 4,6-diamino-5-(tolylthioureido)aminopyrimidine, respectively.

Application

p-Tolyl isothiocyanate has been used:
  • in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione
  • as capping agent to cap the N-terminal ends of polypeptides

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D H Swenson et al.
Chemical research in toxicology, 6(4), 480-485 (1993-07-01)
Methods for direct synthesis of N-(purin-8-yl)arylamines were investigated. N-(Purin-8-yl)arylamines are adducts from reaction of electrophilic metabolites of arylamines with DNA and have not been readily available by direct synthesis. Ability to generate significant quantities of this class of DNA adduct
Thorfinnur Gunnlaugsson et al.
The Journal of organic chemistry, 70(26), 10875-10878 (2005-12-17)
[structure: see text] The synthesis and UV-vis and NMR spectroscopic studies of thiourea-based colorimetric sensors for anions are presented. These sensors can recognize anions through hydrogen binding even in competitive pH-buffered aqueous solutions, giving rise to large color changes that
Polypeptide end-capping using functionalized isocyanates: preparation of pentablock copolymers.
Brzezinska KR, et al.
Macromolecules, 35(8), 2970-2976 (2002)
Karlee L Bamford et al.
Dalton transactions (Cambridge, England : 2003), 49(48), 17571-17577 (2020-11-25)
The synthesis and isolation of the first stable C-B-N-substituted borinium [MesBNiPr2][B(C6F5)4] (2) is described. Compound 2 was shown to react with isothiocyanate and carbodiimides, effecting B-C insertion to afford nitrilium (4) and borenium amidinate salts, respectively. The borinium cation [MesBNiPr2]+
Linhua Huang et al.
Molecules (Basel, Switzerland), 25(11) (2020-06-03)
Due to the side effects of obesity medications, many studies have focused on the natural products used in the daily diet to control weight. Moringa seed pods and leaves are widely used as vegetables or diet supplements due to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service