Skip to Content
Merck
All Photos(1)

Key Documents

189278

Sigma-Aldrich

p-Toluenesulfonyl isocyanate

96%

Synonym(s):

Tosyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NCO
CAS Number:
Molecular Weight:
197.21
Beilstein:
391287
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 100 °C)

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.534 (lit.)

bp

144 °C/10 mmHg (lit.)
275-278 °C (lit.)

density

1.291 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)N=C=O

InChI

1S/C8H7NO3S/c1-7-2-4-8(5-3-7)13(11,12)9-6-10/h2-5H,1H3

InChI key

VLJQDHDVZJXNQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Toluenesulfonyl isocyanate undergoes palladium-catalyzed bis-allylation reaction with allylstannanes and allyl chlorides.

Application

p-Toluenesulfonyl isocyanate has been used as :
  • derivatization reagent in determination of 3-α-hydroxy tibolone in human plasma by LC-MS/MS
  • derivatization reagent in simultaneous quantitative analysis of diethylene glycol and propylene glycol in pharmaceutical products by HPLC
  • as reagent in the preparation of acetylated syn-1,2-diols, oxazolidin-2-ones, 2,3-diamino acids and N-tosylcarbonamides

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

294.8 °F - closed cup

Flash Point(C)

146 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 34, 951-951 (1993)
Tetrahedron Letters, 34, 1867-1867 (1993)
Fabiana Fernandes de Santana e Silva Cardoso et al.
Biomedical chromatography : BMC, 27(11), 1457-1462 (2013-07-03)
A new method was developed for the quantitation of 3-α-hydroxy tibolone, in human plasma, after oral administration of a tablet formulation containing tibolone (2.5 mg). 3-α-Hydroxy tibolone was extracted by a liquid-liquid procedure, using cyproterone acetate as internal standard and chlorobutane
N Solin et al.
Organic letters, 3(6), 909-912 (2001-03-27)
A tandem bis-allylation of p-toluenesulfonyl isocyanate can be achieved by palladium-catalyzed three-component coupling reaction with allylstannanes and allyl chlorides. A high level of regioselectivity can be obtained by the appropriate choice of the allylic substituents. The reaction mechanism and the
Inga Mädge et al.
Food chemistry, 187, 491-498 (2015-05-16)
A general contamination of tea with pyrrolizidine alkaloids (PA) has just become known. Here, we report the application and modification of a new HPLC-ESI-MS/MS sum parameter method to quantitate PA content of herbal teas intended for infants, pregnant and lactating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service