Skip to Content
Merck
All Photos(1)

Key Documents

139742

Sigma-Aldrich

Phenyl isothiocyanate

reagent grade, 98%

Synonym(s):

PITC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NCS
CAS Number:
Molecular Weight:
135.19
Beilstein:
471392
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Assay

98%

form

liquid

refractive index

n20/D 1.6515 (lit.)

bp

218 °C (lit.)

mp

−21 °C (lit.)

solubility

water: insoluble

density

1.132 g/mL at 20 °C (lit.)

functional group

isothiocyanate

SMILES string

S=C=Nc1ccccc1

InChI

1S/C7H5NS/c9-6-8-7-4-2-1-3-5-7/h1-5H

InChI key

QKFJKGMPGYROCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenyl isothiocyanate is an aromatic isothiocyanate. It participates in dehydration reactions of alcohols. It is widely used for synthesis of various biologically important heterocyclic compounds.

Application

Phenyl isothiocyanate may be employed as a derivatization reagent for high-performance liquid chromatographic (HPLC) analysis of various amphetamine derivatives in body fluids for forensic purposes.
Phenyl isothiocyanate (PIT) modified with polyethylene glycol (PEG), can be used as a reagent for PEGylation of protein using human serum albumin as a model protein.
Derivatizing reagent for primary and secondary amines. Used in sequencing peptides by Edman degradation and in amino acid analyses by HPLC.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PEGylation of human serum albumin: reaction of PEG-phenyl-isothiocyanate with protein
Meng F, et al.
Bioconjugate Chemistry, 19(7), 1352-1360 (2008)
Amin Zolali et al.
Combinatorial chemistry & high throughput screening, 17(7), 610-613 (2014-03-19)
An efficient, one-pot and three-component synthesis of biologically important heterocyclic compounds is described from the reaction of primary amines and phenyl isothiocyanate in the presence of acryloyl chloride at room temperature without the need to use any catalyst.
One-pot dehydrations using phenyl isothiocyanate.
Majetich G, et al.
Tetrahedron Letters, 56(23), 3326-3329 (2015)
Determination of phenylisothiocyanate derivatives of amphetamine and its analogues in biological fluids by HPLC-APCI-MS or DAD.
Bogusz MJ, et al.
Journal of Analytical Toxicology, 21(1), 59-69 (1997)
R Gheshlaghi et al.
Analytical biochemistry, 383(1), 93-102 (2008-08-30)
Modified resolution and overall separation factors used to quantify the separation of complex chromatography systems are described. These factors were proven to be applicable to the optimization of amino acid resolution in reverse-phase (RP) HPLC chromatograms. To optimize precolumn derivatization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service