Skip to Content
Merck
All Photos(1)

Key Documents

252182

Sigma-Aldrich

5,10,15,20-Tetraphenyl-21H,23H-porphine copper(II)

Synonym(s):

Copper TPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H28CuN4
CAS Number:
Molecular Weight:
676.27
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

λmax

411 nm
536 nm (2nd)

SMILES string

[Cu]1n2c3ccc2c(-c4ccccc4)c5ccc(n5)c(-c6ccccc6)c7ccc(c(-c8ccccc8)c9ccc(n9)c3-c%10ccccc%10)n17

InChI

1S/C44H28N4.Cu/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36;/h1-28H;/q-2;+2/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;

InChI key

RKTYLMNFRDHKIL-DAJBKUBHSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric spin density distribution in the copper(II) complex of N-confused tetraphenylporphyrin: a multifrequency continuous-wave and pulse EPR study.
George Mitrikas et al.
Angewandte Chemie (International ed. in English), 44(21), 3301-3303 (2005-04-30)
S P Greiner et al.
Journal of biomolecular structure & dynamics, 9(5), 837-851 (1992-04-01)
Proton ENDOR has been observed from frozen solutions (ca. 38K degrees) of copper meso-(4-N-tetra-methylpyridyl)porphyrin (CuTMpyP(4)) complexed with Salmon sperm DNA in water and D2O. Lines from exchangeable protons of the DNA bases have been observed in these ENDOR spectra. Analyses
Ki Young Yeon et al.
Chemical communications (Cambridge, England), 46(30), 5572-5574 (2010-06-26)
Soret band lifetimes of the free-base tetraphenylporphine (H(2)TPP) and Cu(ii) tetraphenylporphine (Cu(II)TPP) at 408 nm have been directly measured with femtosecond (fs) resolution using the fluorescence-upconversion technique for the first time, giving tau = 68 +/- 15 and 63 +/-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service