Skip to Content
Merck
All Photos(1)

Key Documents

252166

Sigma-Aldrich

5,10,15,20-Tetraphenyl-21H,23H-porphine zinc

Synonym(s):

meso-Tetraphenylporphine, TPP, Zinc TPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H28N4Zn
CAS Number:
Molecular Weight:
678.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

λmax

418 nm

OLED Device Performance

ITO/TPD/Alq3:TPP (3%)/Alq3/Mg:Ag

  • Color: red
  • Max. Luminance: 42 V
  • Max. EQE: 0.07 %

SMILES string

[Zn]1n2c3ccc2c(-c4ccccc4)c5ccc(n5)c(-c6ccccc6)c7ccc(c(-c8ccccc8)c9ccc(n9)c3-c%10ccccc%10)n17

InChI

1S/C44H28N4.Zn/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36;/h1-28H;/q-2;+2/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;

InChI key

XPVVGUHKLPZAEN-DAJBKUBHSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V L Schlegel et al.
Analytical chemistry, 63(3), 241-247 (1991-02-01)
The relationship between surface-enhanced resonance Raman scattering (SERRS) intensity and the rate of deposition during silver-island film preparation was examined, using zinc tetraphenylporphine (ZnTPP) as the adsorbate. The effect of the deposition rate on the optical properties of the films
H Kolarova et al.
Toxicology in vitro : an international journal published in association with BIBRA, 19(7), 971-974 (2005-08-03)
The basis of photodynamic therapy (PDT) is the phototoxicity resulting from co-action of light, sensitizer and oxygen. In this study we demonstrate in vitro phototoxicity measurement on G361 cell lines using ZnTPPS(4) sensitizer bound to cyclodextrin hpbetaCD. We have proved
Martin Kullmann et al.
Physical chemistry chemical physics : PCCP, 14(22), 8038-8050 (2012-05-03)
We report on a comprehensive transient absorption study with β,β'-linked bis[tetraphenylporphyrinato-zinc(II)] and its corresponding monomer, covering the ultrafast dynamics from femtoseconds up to several microseconds. By exciting these porphyrins either to their first (S(1)) or second (S(2)) electronically excited states
Vijaykumar S Ijeri et al.
Langmuir : the ACS journal of surfaces and colloids, 24(3), 1070-1079 (2008-01-05)
Active spatial and temporal modulation of domains of singlet oxygen activity is demonstrated using electrochemical tools. Using singlet oxygen microscopy in photosensitized experiments, it is shown that singlet oxygen concentrations around an ultramicroelectrode can be controlled by applying a bias
Suyi Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 88, 137-143 (2012-01-10)
The molecular interactions of 5,10,15,20-tetraphenylporphine zinc (ZnTPP) and 5,10,15,20-tetraphenyl-21H,23H-porphine cobalt(II) (CoTPP) with dimethyl methylphosphonate (DMMP) have been investigated by absorption/absorption difference spectroscopy. The interactions between the metalloporphyrins and DMMP change the absorbance characteristics of the porphyrins resulted from the formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service