Skip to Content
Merck
All Photos(1)

Key Documents

258423

Sigma-Aldrich

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine zinc(II)

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H44N4Zn
CAS Number:
Molecular Weight:
598.15
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

λmax

400 nm

SMILES string

CCc1c(CC)c2cc3c(CC)c(CC)c4cc5nc(cc6c(CC)c(CC)c(cc1n2)n6[Zn]n34)c(CC)c5CC

InChI

1S/C36H44N4.Zn/c1-9-21-22(10-2)30-18-32-25(13-5)26(14-6)34(39-32)20-36-28(16-8)27(15-7)35(40-36)19-33-24(12-4)23(11-3)31(38-33)17-29(21)37-30;/h17-20H,9-16H2,1-8H3;/q-2;+2/b29-17-,30-18-,31-17-,32-18-,33-19-,34-20-,35-19-,36-20-;

InChI key

VVUVWOLOUOYXOI-XTPDIVBZSA-N

General description

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine zinc(II) ) is formed by metalating 2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine with zinc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service