Skip to Content
Merck
All Photos(3)

Key Documents

250899

Sigma-Aldrich

1,5-Naphthalenedisulfonic acid disodium salt hydrate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(SO3Na)2 · xH2O
CAS Number:
Molecular Weight:
332.26 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

functional group

sulfonic acid

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)c1cccc2c(cccc12)S([O-])(=O)=O

InChI

1S/C10H8O6S2.2Na/c11-17(12,13)9-5-1-3-7-8(9)4-2-6-10(7)18(14,15)16;;/h1-6H,(H,11,12,13)(H,14,15,16);;/q;2*+1/p-2

InChI key

YGSZNSDQUQYJCY-UHFFFAOYSA-L

General description

The liposomal entrapment of 1,5-naphthalenedisulfonic acid disodium salt in phospholipid vesicles has been studied. Doubly substituted aromatic dianions of 1,5-naphthalenedisulfonic acid disodium salt has been investigated by collision-induced dissociation, along with infrared multiple photon dissociation/detachment techniques.

Other Notes

Contains sodium chloride

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Shaun Ard et al.
The Journal of chemical physics, 132(9), 094301-094301 (2010-03-10)
Collision-induced dissociation (CID), along with infrared multiple photon dissociation/detachment (IRMPD) techniques, is utilized to study a series of doubly substituted aromatic dianions containing sulfonate and carboxylate functionalities (1,2- and 1,3-benzenedisulfonate, 1,5-naphthalenedisulfonate, 2,6-naphthalenedisulfonate, 4-sulfobenzoate, 2,6-naphthalenedicarboxylate, and terephthalate dianions). The molecules were
H C Chang et al.
Journal of pharmaceutical sciences, 83(7), 1043-1046 (1994-07-01)
The liposomal entrapment of suramin and similar compounds in phospholipid vesicles was examined. For dipalmitoylphosphatidylcholine (DPPC) liposomes, entrapment percentages ranged from 25 to 65% with 3-25 mM phospholipid for aqueous solutions containing 0.07 mM of suramin. Incorporation of 30-50 mol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service