218766
4-Aminotoluene-3-sulfonic acid
99%
Synonym(s):
2-Amino-5-methylbenzenesulfonic acid, 4-Methylaniline-2-sulfonic acid
About This Item
Recommended Products
Assay
99%
form
solid
solubility
water: soluble 6 g/L at 20 °C
functional group
sulfonic acid
SMILES string
Cc1ccc(N)c(c1)S(O)(=O)=O
InChI
1S/C7H9NO3S/c1-5-2-3-6(8)7(4-5)12(9,10)11/h2-4H,8H2,1H3,(H,9,10,11)
InChI key
LTPSRQRIPCVMKQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service