Skip to Content
Merck
All Photos(3)

Key Documents

232114

Sigma-Aldrich

Platinum on alumina

greener alternative

extent of labeling: 1 wt. % loading, 3.2 mm, pellets

Synonym(s):

Pt/Al2O3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

pellets

Quality Level

reaction suitability

core: platinum
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

1 wt. % loading

particle size

3.2 mm

greener alternative category

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Platinum on alumina (Pt/Al2O3) (1 wt. % loading, 3.2 mm, pellets) can be used as a catalyst for a variety of applications, such as:
  • in the hydrodechlorination of chlorobenzenes
  • oxidation, reduction, and decomposition
  • in methanol steam reforming
  • hydrocarbon combustion

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paquette, L. A.; Miyahara, Y.; Doecke, C. W.
Journal of the American Chemical Society, 1716, 1716-1716 (1986)
Preparation of Platinum-Supported Alumina Lewis Superacid Catalyst Using a CVD Method
Ayame A, et al.
Studies in Surface Science and Catalysis, 90, 257-262 (1994)
Kinetics of o-Xylene Combustion over a Pt/Alumina Catalyst
Manta CM, et al.
Chemical Engineering & Technology, 35(12), 2147-2154 (2012)
Low-temperature hydrodechlorination of chlorobenzenes on platinum-supported alumina catalysts
Hashimoto Y, et al.
Applied Catalysis A: General, 250(12), 247-254 (2003)
Fraenkel, G.; Gallucci, J.; Rosenzweig, H. S.
The Journal of Organic Chemistry, 54, 677-677 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service