Skip to Content
Merck
All Photos(3)

Key Documents

208779

Sigma-Aldrich

Nickel on silica/alumina

greener alternative

extent of labeling: ~65 wt. % loading, powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

purified by

reduction and stabilization

reaction suitability

reagent type: catalyst
core: nickel

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

~65 wt. % loading

surface area

~175 m2/g

greener alternative category

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

Application

Nickel on silica/alumina can be used as an efficient catalyst in a variety of applications such as:
  • in plastic waste pyrolysis for the production of fuel
  • to study the supercritical water reforming of glycerol
  • in the supercritical water reforming of acetic acid for the production of hydrogen

Other Notes

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1A Inhalation - Carc. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Turnover rates for the supercritical water reforming of glycerol on supported Ni and Ru catalysts
Ortiz FJG, et al.
Fuel: The Science and Technology of Fuel and Energy, 180(21), 417-423 (2016)
Catalytic pyrolysis of municipal plastic waste to fuel with nickel-loaded silica-alumina catalysts
Wang J-L and Wang L-L
Energy Sources, Part A: Recovery, Utilization, and Environmental Effects, 33(21), 1940-1948 (2011)
Hydrogen production from supercritical water reforming of acetic acid, acetol, 1-butanol and glucose over Ni-based catalyst
Ortiz FJG and Campanario FJ
Journal of Supercritical Fluids, 138(21), 259-270 (2018)
Supercritical water reforming of glycerol: Performance of Ru and Ni catalysts on Al2O3 support
Ortiz FJG, et al.
Energy, 96(21), 561-568 (2016)

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service