Skip to Content
Merck
All Photos(1)

Key Documents

I9805

Sigma-Aldrich

1-Iodo-4-nitrobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4NO2
CAS Number:
Molecular Weight:
249.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

bp

289 °C/772 mmHg (lit.)

mp

171-173 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(I)cc1

InChI

1S/C6H4INO2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H

InChI key

SCCCFNJTCDSLCY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loai Basheer et al.
Food chemistry, 237, 895-903 (2017-08-03)
Inhibition of cytochrome P450 3A4 (CYP3A4), the major drug metabolizing enzyme, by dietary compounds has recently attracted increased attention. Evaluating the potency of the many known inhibitory compounds is a tedious and time consuming task, yet it can be achieved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service