Skip to Content
Merck
All Photos(1)

Key Documents

222771

Sigma-Aldrich

4-(Methylthio)benzaldehyde

95%

Synonym(s):

4-(Methylmercapto)benzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SC6H4CHO
CAS Number:
Molecular Weight:
152.21
Beilstein:
636158
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.646 (lit.)

bp

86-90 °C/1 mmHg (lit.)

density

1.144 g/mL at 25 °C (lit.)

functional group

aldehyde
thioether

SMILES string

[H]C(=O)c1ccc(SC)cc1

InChI

1S/C8H8OS/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3

InChI key

QRVYABWJVXXOTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Shin et al.
Bioorganic & medicinal chemistry letters, 11(2), 165-168 (2001-02-24)
A series of 2,2-dimethyl-5-[4-(methylsulfonyl)phenyl]-4-phenyl-3(2H)furanones was prepared and evaluated for their ability to inhibit cyclo-oxygenase-2 (COX-2).
Thomas Keller et al.
Journal of biophotonics, 13(6), e201960126-e201960126 (2020-01-21)
The characteristic vibrational spectroscopic fingerprint of Raman reporter molecules adsorbed on noble metal nanoparticles is employed for the identification of target proteins by the corresponding surface-enhanced Raman scattering (SERS) nanotag-labeled antibodies. Here, we present the modular synthesis of thiolated polyenes
Coraline Simon et al.
Toxicology in vitro : an international journal published in association with BIBRA, 37, 121-133 (2016-10-19)
Endocrine activity of 65 compounds migrating from polycarbonate replacement plastic baby bottles was assessed using in vitro cell based assays (reporter gene assays) involving 7 nuclear receptors, i.e. human steroid hormones receptors (oestrogen, androgen, progesterone and glucocorticoid receptors), human thyroid
Matthias Onghena et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 31(12), 2090-2102 (2014-11-20)
In 2011, the European Union prohibited the production of polycarbonate (PC) baby bottles due to the toxic effects of the PC monomer bisphenol-A. Therefore, baby bottles made of alternative materials, e.g. polypropylene (PP) or polyethersulphone (PES), are currently marketed. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service