Skip to Content
Merck
All Photos(1)

Key Documents

274240

Sigma-Aldrich

2-Aminothiophenol

99%

Synonym(s):

2-Aminobenzenethiol, 2-Aminophenyl mercaptan, 2-Mercaptoaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4SH
CAS Number:
Molecular Weight:
125.19
Beilstein:
606076
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.642 (lit.)

bp

70-72 °C/0.2 mmHg (lit.)

mp

16-20 °C (lit.)

density

1.17 g/mL at 25 °C (lit.)

SMILES string

Nc1ccccc1S

InChI

1S/C6H7NS/c7-5-3-1-2-4-6(5)8/h1-4,8H,7H2

InChI key

VRVRGVPWCUEOGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Aminothiophenol is an organosulfur compound used in the synthesis of urea derivatives.

Application

2-Aminothiophenol was used in the synthesis of 1,5-benzothiazepines derivatives by reacting with 1,3-diphenylpropenone derivatives and using aluminosilicate solid catalysts. It was also used in the synthesis of benzothiazole and 3-(benzothiazol-2-yl)coumarin derivatives.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vikas S Patil et al.
Journal of fluorescence, 23(5), 1019-1029 (2013-05-18)
Novel ESIPT inspired benzimidazole, benzoxazole and benzothiazole were synthesized from 2,4-dihydroxy benzoic acid and 1,2-phenelenediamine, 2-aminophenol, and 2-aminothiophenol respectively. The synthesized 2-(2',4'-dihydroxyphenyl) benzimidazole, benzoxazole and benzothiazole are fluorescent and the emission characteristic are very sensitive to the micro-environment. They show
Maria J Climent et al.
ChemSusChem, 7(4), 1177-1185 (2014-03-13)
1,5-Benzothiazepines derivatives were obtained first by starting from 1,3-diphenylpropenone derivatives (chalcones) and 2-aminothiophenol by using aluminosilicate solid catalysts. However, diffusional limitations and the strong adsorption of products on the catalyst are deleterious for catalyst activity and life. Then a structured
Chemoselective synthesis, X-ray characterization and DFT studies of new organic single crystal: S-(2-aminophenyl) cyclohexylcarbamothioate
Dougan S, et al.
Journal of Molecular Structure, 1204, 127499-127499 (2020)
Tianzhi Yu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 108, 274-279 (2013-03-19)
Two different types of fluorescent materials containing benzothiazolyl moiety, 2-(benzothiazol-2-yl)phenol derivatives and 3-(benzothiazol-2-yl)coumarin derivatives, were synthesized synchronously using ethyl cyanoacetate, appropriate aromatic aldehyde and 2-aminothiophenol as the starting materials under the catalysis of benzoic acid by one-pot reaction. This method
William P Morrow et al.
Archives of biochemistry and biophysics, 631, 66-74 (2017-08-23)
Thiol dioxygenases are non-heme mononuclear iron enzymes that catalyze the O

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service