Skip to Content
Merck
All Photos(1)

Key Documents

211265

Sigma-Aldrich

4-Phenoxybenzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OC6H4CHO
CAS Number:
Molecular Weight:
198.22
Beilstein:
1947841
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.611 (lit.)

bp

185 °C/14 mmHg (lit.)

mp

24-25 °C (lit.)

density

1.132 g/mL at 25 °C (lit.)

functional group

aldehyde
phenoxy

SMILES string

O=Cc1ccc(Oc2ccccc2)cc1

InChI

1S/C13H10O2/c14-10-11-6-8-13(9-7-11)15-12-4-2-1-3-5-12/h1-10H

InChI key

QWLHJVDRPZNVBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Phenoxybenzaldehyde was used in the synthesis of:
  • spirodiketopiperazine derivatives
  • benzoxazoles

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rena Nishizawa et al.
Bioorganic & medicinal chemistry letters, 17(3), 727-731 (2006-11-23)
Hydroxylated derivatives were designed and synthesized based on the information of oxidative metabolites. Compounds derived from beta-substituted (2R,3R)-2-amino-3-hydroxypropionic acid showed improved inhibitory activities against the binding of MIP-1alpha to human CCR5, compared with the non-hydroxylated derivatives and the other isomers.
Kelly J McClure et al.
Bioorganic & medicinal chemistry letters, 16(7), 1924-1928 (2006-01-31)
In a recent paper, [Arienti, K. L.; Brunmark, A.; Axe, F. U.; McClure, K. M.; Lee, A.; Blevitt, J.; Neff, D. K.; Huang, L.; Crawford, S.; Chennagiri, R. P.; Karlsson, L.; Brietenbucher, J. G. J. Med. Chem.2005, 48, 1873], we
Vikas N Telvekar et al.
Bioorganic & medicinal chemistry letters, 22(1), 649-652 (2011-11-15)
A series of structurally novel, substituted 2-(2-(4-aryloxybenzylidene) hydrazinyl)benzothiazole derivatives incorporating 2-hydrazinyl benzothiazole and 4-(aryloxy)benzaldehyde were designed and synthesized using molecular hybridization approach. All the synthesized compounds exhibited promising activity (MIC 1.5-29.00μg/ml) against Mycobacteriumtuberculosis H37Rv strains of using REMA. Five of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service