Skip to Content
Merck
All Photos(1)

Key Documents

220159

Sigma-Aldrich

2,3-Dimethyl-2-butene

≥99%

Synonym(s):

Tetramethylethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=C(CH3)2
CAS Number:
Molecular Weight:
84.16
Beilstein:
1361357
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

215 mmHg ( 37.7 °C)

Quality Level

Assay

≥99%

form

liquid

autoignition temp.

754 °F

refractive index

n20/D 1.412 (lit.)

bp

73 °C (lit.)

mp

−75 °C (lit.)

density

0.708 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C\C(C)=C(\C)C

InChI

1S/C6H12/c1-5(2)6(3)4/h1-4H3

InChI key

WGLLSSPDPJPLOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Dimethyl-2-butene undergoes ozonolysis in dark to yield hydroxyl radical. Reaction of ozone with 2,3-dimethyl 2-butene (DMB) has been investigated using a flow-tube interfaced to UV photoelectron spectrometer. DMB forms adduct with thianthrene cation radical tetrafluoroborate at 0°C and -15°C.

Application

2,3-Dimethyl-2-butene was employed as substrate in photoinduced molecular transformations involving 2-hydroxy-1,4-naphthoquinones.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

17.6 °F - closed cup

Flash Point(C)

-8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 58, 4614-4614 (1993)
Bing-Jun Zhao et al.
The Journal of organic chemistry, 71(10), 3737-3742 (2006-05-06)
Thianthrene cation radical tetrafluoroborate (Th*+ BF4-) has been found to add to 2,3-dimethyl-2-butene (DMB) at 0 degrees C and -15 degrees C. The adduct, 2,3-dimethyl-2,3-(5,10-thianthreniumdiyl)butane ditetrafluoroborate (12), was isolated at -15 degrees C, and its 1H NMR spectrum was recorded
Maryline Pflieger et al.
Environmental science & technology, 47(12), 6239-6246 (2013-05-15)
In order to investigate the heterogeneous oxidation kinetics of the herbicide terbuthylazine (TERB), a stable and reproducible generation system of "dark" hydroxyl radical in the gas phase was developed and optimized using a PTR-MS. TERB was adsorbed on silica particles
Andrew T Lambe et al.
Environmental science & technology, 41(7), 2357-2363 (2007-04-19)
We present a novel method for continuous, stable OH radical production for use in smog chamber studies, especially those focused on organic aerosol aging. Our source produces OH radicals from the reaction of 2,3-dimethyl-2-butene and ozone and is unique as
N Haga et al.
Photochemistry and photobiology, 61(6), 557-562 (1995-06-01)
The mechanism of photocycloaddition of 2'-deoxyuridine (1a) and thymidine (1b) to 2,3-dimethyl-2-butene (Bu) in acetonitrile by UV irradiation has been studied. The reciprocal quantum yield for the cycloaddition increased linearly with reciprocal concentrations of Bu in acetonitrile to give limiting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service