Skip to Content
Merck
All Photos(1)

Key Documents

190403

Sigma-Aldrich

2,3-Dimethyl-1-butene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC(CH3)=CH2
CAS Number:
Molecular Weight:
84.16
Beilstein:
505982
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

412 mmHg ( 37.7 °C)

Quality Level

Assay

97%

form

liquid

autoignition temp.

680 °F

refractive index

n20/D 1.389 (lit.)

bp

56 °C (lit.)

mp

−158 °C (lit.)

density

0.68 g/mL at 25 °C (lit.)

SMILES string

CC(C)C(C)=C

InChI

1S/C6H12/c1-5(2)6(3)4/h6H,1H2,2-4H3

InChI key

OWWIWYDDISJUMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gas phase reaction of 2,3-dimethyl-1-butene with the OH radical has been investigated in the presence of NO.

Application

2,3-Dimethyl-1-butene was used to investigate the mechanism of the sulfur vulcanization of rubber.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sulfur Vulcanization of Simple Model Olefins Part III: Vulcanization of 2, 3-Dimethyl-2-Butene in the Presence of Different Metal Complexes.
Versloot P, et al.
Rubber Chemistry and Technology, 67(2), 263-279 (1994)
Products of the Gas-Phase Reactions of a Series of 1-Alkenes and 1-Methylcyclohexene with the OH Radical in the Presence of NO.
R Atkinson et al.
Environmental science & technology, 29(6), 1674-1680 (1995-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service