119059
3,3-Dimethyl-1-butene
≥97% (GC)
Synonym(s):
Neohexene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)3CCH=CH2
CAS Number:
Molecular Weight:
84.16
Beilstein:
1697101
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
>1 (vs air)
Quality Level
vapor pressure
6.96 psi ( 20 °C)
Assay
≥97% (GC)
refractive index
n20/D 1.376 (lit.)
bp
41 °C (lit.)
mp
−115 °C (lit.)
density
0.653 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CC(C)(C)C=C
InChI
1S/C6H12/c1-5-6(2,3)4/h5H,1H2,2-4H3
InChI key
PKXHXOTZMFCXSH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
3,3-Dimethyl-1-butene oxidizes selectively to corresponding epoxide by chlorine promoted Ag(111) oxygen adatoms. It undergoes copolymerization with ethylene using a chain-walking Pd-diimine catalyst.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Asp. Tox. 1 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
17.6 °F - closed cup
Flash Point(C)
-8 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Peng Xiang et al.
Macromolecular rapid communications, 31(12), 1083-1089 (2011-05-19)
In this Communication, the copolymerization of ethylene with a sterically hindered α-olefin comonomer, γ-trisubstituted 3,3-dimethyl-1-butene (DMB), using a chain-walking Pd-diimine catalyst, [(ArNC(Me)(Me)CNAr)Pd(CH(3) )(NCMe)]SbF(6) (Ar2,6-(iPr)(2) C(6) H(3) ) (1) is reported. In spite of its high steric bulkiness in the close
Molecular mechanism of alkene epoxidation: A model study with 3, 3-dimethyl-1-butene on Ag (111).
Mukoid C, et al.
Catalysis Letters, 4(1), 57-61 (1990)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service