Skip to Content
Merck
All Photos(2)

Key Documents

366706

Sigma-Aldrich

1-Iodo-3,5-bis(trifluoromethyl)benzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C6H3I
CAS Number:
Molecular Weight:
340.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.463 (lit.)

bp

59-61 °C/10 mmHg (lit.)

density

1.919 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1cc(I)cc(c1)C(F)(F)F

InChI

1S/C8H3F6I/c9-7(10,11)4-1-5(8(12,13)14)3-6(15)2-4/h1-3H

InChI key

VDPIZIZDKPFXLI-UHFFFAOYSA-N

Related Categories

General description

1-Iodo-3,5-bis(trifluoromethyl)benzene is a halogenated hydrocarbon.

Application

1-Iodo-3,5-bis(trifluoromethyl)benzene may be used in the preparation of [bis(trifluoroacetoxy)iodo]perfluoroalkanes. It may be used to investigate the fragmentation reactions of (E)- and (Z)-2-methylbuten-1-yl(aryl)iodonium triflates (aryl = C6H5-, 4-(CF3)C6H4, 3,5-(CF3)2C6H4-).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksandra A Zagulyaeva et al.
The Journal of organic chemistry, 75(6), 2119-2122 (2010-02-19)
[Bis(trifluoroacetoxy)iodo]perfluoroalkanes C(n)F(2n+1)I(OCOCF(3))(2) (n = 4, 6, 8, 10, 12) can be conveniently prepared by the oxidation of the corresponding perfluoroalkyl iodides with Oxone in trifluoroacetic acid at room temperature and subsequently converted to the stable [hydroxy(tosyloxy)iodo]perfluoroalkanes, C(n)F(2n+1)I(OH)OTs, by treatment with
Fragmentations of (E)-and (Z)-isomers of 2-methylbuten-1-yl (aryl) iodonium triflates: competing mechanisms for enol triflate formation.
Hinkle RJ.
ARKIVOC (Gainesville, FL, United States), 6, 201-212 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service