Skip to Content
Merck
All Photos(2)

Key Documents

212490

Sigma-Aldrich

Direct Red 23

Dye content 30 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H25N7Na2O10S2
CAS Number:
Molecular Weight:
813.72
Colour Index Number:
29160
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 30%

technique(s)

microbe id | staining: suitable

λmax

507 nm

ε (extinction coefficient)

≥15000 at 500-516 nm in water at 0.04 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].CC(=O)Nc1ccc(cc1)N=Nc2cc3c(O)cc(NC(=O)Nc4ccc5c(O)c(N=Nc6ccccc6)c(cc5c4)S([O-])(=O)=O)cc3cc2S([O-])(=O)=O

InChI

1S/C35H27N7O10S2.2Na/c1-19(43)36-22-7-9-24(10-8-22)40-42-32-30(54(50,51)52)18-21-16-26(12-14-28(21)34(32)45)38-35(46)37-25-11-13-27-20(15-25)17-29(53(47,48)49)31(33(27)44)41-39-23-5-3-2-4-6-23;;/h2-18,44-45H,1H3,(H,36,43)(H2,37,38,46)(H,47,48,49)(H,50,51,52);;/q;2*+1/p-2

InChI key

NLMHXPDMNXMQBY-UHFFFAOYSA-L

General description

Direct Red 23 consists of a diazo group and it is toxic and carcinogenic in nature.

Application

Direct Red 23 has been used to stain water-hydrated seeds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Removal of Direct Red 23 from aqueous solution using corn stalks: Isotherms, kinetics and thermodynamic studies
Fathi MR
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 364-372 (2015)
Extensive Natural Variation in Arabidopsis Seed Mucilage Structure
Voiniciuc C
Frontiers in Plant Science, 7 (2016)
Amina Yaqoob et al.
PloS one, 15(3), e0230519-e0230519 (2020-03-19)
Promoters are specified segments of DNA that lead to the initiation of transcription of a specific gene. The designing of a gene cassette for plant transformation is significantly dependent upon the specificity of a promoter. Constitutive Cauliflower mosaic virus promoter
Dongbo Shi et al.
Development (Cambridge, England), 146(1) (2019-01-11)
A reduced rate of stem cell division is considered a widespread feature which ensures the integrity of genetic information during somatic development of plants and animals. Radial growth of plant shoots and roots is a stem cell-driven process that is
Julien Moukhtar et al.
PLoS computational biology, 15(2), e1006771-e1006771 (2019-02-12)
Plant tissue architecture and organ morphogenesis rely on the proper orientation of cell divisions. Previous attempts to predict division planes from cell geometry in plants mostly focused on 2D symmetric divisions. Using the stereotyped division patterns of Arabidopsis thaliana early

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service