Skip to Content
Merck
All Photos(2)

Key Documents

F5005

Sigma-Aldrich

3-Fluorobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CHO
CAS Number:
Molecular Weight:
124.11
Beilstein:
970178
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.518 (lit.)

bp

66-68 °C/20 mmHg (lit.)

density

1.17 g/mL at 25 °C (lit.)

SMILES string

Fc1cccc(C=O)c1

InChI

1S/C7H5FO/c8-7-3-1-2-6(4-7)5-9/h1-5H

InChI key

PIKNVEVCWAAOMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - DIN 51755 Part 1

Flash Point(C)

56 °C - DIN 51755 Part 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Belén Ruiz-Muelle et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 22(7), 2427-2439 (2016-01-21)
The synthesis and structural characterization of allenyl titanocene(IV) [TiClCp2 (CH=C=CH2 )] 3 and propargyl titanocene(IV) [TiClCp2 (CH2 -C≡C-(CH2 )4 CH3 )] 9 have been described for the first time. Advanced NMR methods including diffusion NMR methods (diffusion pulsed field gradient
Syed Saad Hussain et al.
Molecular and cellular biochemistry, 471(1-2), 29-39 (2020-05-31)
P53 plays an important role in maintaining genetic stability and development of resistance against tumors. Dysregulation of P53 gene is one of the key factors contributing to the etiology of neuroblastoma which causes cells to evade apoptosis. Activating P53 pathway
Kateřina Štulíková et al.
Molecules (Basel, Switzerland), 25(10) (2020-05-28)
Due to the increasing popularity of unfiltered beer, new methods for its preservation are needed. High-pressure processing (HPP) was applied as a final treatment of packed beer in order to assure storage stability and to retain the desired product quality.
Bin-Yu Wen et al.
Journal of Asian natural products research, 21(7), 702-715 (2019-01-01)
Desmosdumotin C (Des C), a natural product isolated from the roots of Desmos dumosus, has shown good antitumor activity. A three dimensional quantitative structure-activity relationship (QSAR) study using the comparative molecular field analysis (CoMFA) method was performed on 32 Des
Adnan Ashraf et al.
European journal of medicinal chemistry, 159, 282-291 (2018-10-09)
Chalcones and 1,2-benzothiazines are two important classes of bioactive compounds, each scaffold endowed with diverse pharmacological activities. Combining both of these pharmacophores in a single molecule was aimed to yield multi-modal agents. Herein, we report a series of hybrid compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service