Skip to Content
Merck
All Photos(3)

Key Documents

206199

Sigma-Aldrich

Ruthenium on alumina

greener alternative

extent of labeling: 0.5 wt. % loading, pellets, 3.2 mm

Synonym(s):

Ru

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ru
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

pellets

Quality Level

reaction suitability

core: ruthenium
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

0.5 wt. % loading

particle size

3.2 mm

greener alternative category

SMILES string

[Ru]

InChI

1S/Ru

InChI key

KJTLSVCANCCWHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Ruthenium on alumina (Ru/Al2O3) (0.5 wt. % loading, pellets, 3.2 mm) can be used as a catalyst for the production of methane via hydrogenation of carbon dioxide. It can also be used as a catalyst in the reduction of CO2.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methanation of carbon dioxide on Ru/Al2O3: catalytic activity and infrared study
Garbarino G, et al.
Catalysis Today, 277(17), 21-28 (2016)
Study of catalytic hydrogenation of N-ethylcarbazole over ruthenium catalyst
Wan C, et al.
International Journal of Hydrogen Energy, 37(17), 13092-13096 (2012)
Hydrogen and methane selectivity during alkaline supercritical water gasification of biomass with ruthenium-alumina catalyst
Onwudili JA and Williams PT
Applied Catalysis. B, Environmental, 132, 70-79 (2013)
CO2 reduction on supported Ru/Al2O3 catalysts: cluster size dependence of product selectivity
Kwak JH, et al.
ACS Catalysis, 3(11), 2449-2455 (2013)

Articles

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service