Skip to Content
Merck
All Photos(2)

Key Documents

683124

Sigma-Aldrich

Palladium(II) acetate

Umicore, 46.6-49.5% palladium (Pd) basis

Synonym(s):

Pd(OAc)2, [Pd(OAc)2]3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(OCOCH3)2
CAS Number:
Molecular Weight:
224.51
Beilstein:
6086766
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

concentration

46.6-49.5% Pd

mp

216.3-223.7 °C (dec.)

SMILES string

CC(O[Pd]OC(C)=O)=O

InChI

1S/2C2H4O2.Pd/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

YJVFFLUZDVXJQI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinyue Ding et al.
Nature communications, 5, 5474-5474 (2014-11-19)
Non-aromatic nitrogen- and oxygen-containing heterocycles such as piperidines and pyrans are prevalent components of natural products and pharmaceutical drugs. Although it has been a workhorse as a synthetic method for assembling unsaturated sp(2)-hybridized substrates, transition metal-catalysed cross-coupling chemistry is traditionally

Articles

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service