Skip to Content
Merck
All Photos(3)

Key Documents

203645

Sigma-Aldrich

Lithium fluoride

greener alternative

powder, <100 μm, ≥99.98% trace metals basis

Synonym(s):

Fluorolithium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiF
CAS Number:
Molecular Weight:
25.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.98% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤200.0 ppm Trace Metal Analysis

particle size

<100 μm

bp

1673 °C/1 atm (lit.)

mp

845 °C (lit.)

solubility

aqueous acid: slightly soluble(lit.)

density

2.64 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Li+].[F-]

InChI

1S/FH.Li/h1H;/q;+1/p-1

InChI key

PQXKHYXIUOZZFA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Lithium fluoride can be used as an electron injection layer in the fabrication of electroluminescent devices.

It can be used as an electrolyte additive to protect the lithium metal electrode, which improves the stability and reversibility of the battery. The partially soluble LiF forms a thin coating over the surface of lithium and stabilizes the SEI layer.

It can also be used as catalyst support in the preparation of carbon nanofibers by a catalytic chemical vapor deposition method.

Features and Benefits

  • High mechanical strength
  • Wide electrochemical stability window
  • Low calculated barriers toLi diffusion

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asa Carlsson Tedgren et al.
Medical physics, 38(10), 5539-5550 (2011-10-14)
High energy photon beams are used in calibrating dosimeters for use in brachytherapy since absorbed dose to water can be determined accurately and with traceability to primary standards in such beams, using calibrated ion chambers and standard dosimetry protocols. For
Chilin Li et al.
Nano letters, 12(3), 1241-1246 (2012-02-07)
The thermodynamically required redistribution of ions at given interfaces is being paid increased attention. The present investigation of the contact LiF/TiO(2) offers a highly worthwhile example, as the redistribution processes can be predicted and verified. It consists in Li ion
E Cruz-Zaragoza et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 69(10), 1369-1373 (2011-06-21)
The influence of heating rate on the thermoluminescence (TL) property of LiF:Mg,Cu,P+PTFE was analyzed. The activation energy and the frequency factor as a function of the heating rate were determined. The kinetic parameters and their dependence on the heating rate
Viktor Myroshnychenko et al.
Optics express, 20(10), 10879-10887 (2012-05-09)
The availability of macroscopic, nearly periodic structures known as eutectics opens a new path for controlling light at wavelength scales determined by the geometrical parameters of these materials and the intrinsic properties of their component phases. Here, we analyze the
Basile F E Curchod et al.
Chimia, 67(4), 218-221 (2013-08-24)
The implementation of local control theory using nonadiabatic molecular dynamics within the framework of linear-response time-dependent density functional theory is discussed. The method is applied to study the photoexcitation of lithium fluoride, for which we demonstrate that this approach can

Articles

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service