Skip to Content
Merck
All Photos(3)

Key Documents

655201

Sigma-Aldrich

PEDOT:PSS

greener alternative

high-conductivity grade, 3.0-4.0% aqueous dispersion

Synonym(s):

PEDOT:PSS, Poly(2,3-dihydrothieno-1,4-dioxin)-poly(styrenesulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

product name

Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate), 3.0-4.0% in H2O, high-conductivity grade

grade

high-conductivity grade

Quality Level

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

3.0-4.0% in H2O

resistance

1500 Ω/sq, 4 point probe measurement of dried coating based on initial 6μm wet thickness.
500 Ω/sq, 4 point probe measurement of dried coating based on initial 18μm wet thickness.

pH

1.5-2.5 (25 °C, dried coatings)

conductivity

>200 S/cm

viscosity

10-30 cP(20 °C)

density

1.011 g/cm3 (dried coatings)

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate) (PEDOT:PSS) is an organic semiconductor prepared by doping cationic poly(3,4-ethylenedioxythiophene) and poly(4-styrenesulfonate) anion. Its high electrical conductivity and good oxidation resistance make it suitable for electromagnetic shielding and noise suppression. PEDOT:PSS based polymeric films have a high transparency throughout the visible light spectrum and even in near IR and near UV regions, with virtually 100% absorption from 900-2000 nm. PEDOT provides the conduction properties and PSS forms a hydrated colloidal solution.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. This product is used in energy conversion and storage, thus has been enhanced for energy efficiency. Click here for more information.

Application

Electrical conductivity measurements herewith reported were on a film deposited by spin-coating on a clean glass, then dried (130 °C for 15 minutes on a hotplate). The layer thickness was determined by scratching the layer and measuring the profile/height of the scratch by a stylus profilometer. Electrodes for the measurement were by evaporating metal contacts (four-point probes).
PEDOT:PSS is an intrinsically conductive polymer (ICP) that can be coated on various substrates and nanostructures like fullerenes (C60) to form composites with high electrochemical properties for applications like low-cost printed electronics, optoelectronics, and polymeric solar cells. It can be used as a conductive hydrogel with polyethylene glycol-diacrylate (PEG-DA) for potential applications in tissue engineering. PEDOT:PSS also finds use in other organic electronic applications like organic thin film transistors (OTFTs) and dye sensitized solar cells (DSSCs).
Ready-to-use high conductivity coating formulation.
Virtually 100% absorption from 900-2,000 nm. No absorption maximum from 400-800 nm. Conductive polymer blend.

Packaging

Packaged in poly bottles

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanically robust, photopatternable conductive hydrogel composites.
Pal R, et al.
Reactive and Functional Polymers, 120(5), 66-73 (2017)
Dye sensitized solar cells (DSSCs) based on modified iron phthalocyanine nanostructured TiO2 electrode and PEDOT: PSS counter electrode.
Balraju P, et al.
Synthetic Metals, 159(13), 1325-1331 (2009)
Fine patterning of glycerol-doped PEDOT: PSS on hydrophobic PVP dielectric with ink jet for source and drain electrode of OTFTs
Lee M, et al.
Organic Electronics, 11(5}, 854-859 (2010)
New Conducting and Semiconducting Polymers for Organic Photovoltaics.
Sapp S and Luebben S
MRS Online Proceedings Library, 1270(4), 261-266 (2010)
Junhwan Jang et al.
Nanomaterials (Basel, Switzerland), 10(12) (2020-12-03)
Graphene oxide (GO)-cysteamine-Ag nanoparticles (GCA)-silver nanowire (AgNW) fabricated by depositing GCA over sprayed AgNWs on PET films were proposed for transparent and flexible electrodes, and their optical, electrical, and mechanical properties were analyzed by energy-dispersive X-ray spectroscopy, Fourier-transform infrared spectroscopy

Articles

In the field of organic printable electronics, such as OLEDs and organic photovoltaics (OPVs), improved organic conducting and semiconducting materials are needed. The progress in two fields is reviewed in this article.

In the field of organic printable electronics, such as OLEDs and organic photovoltaics (OPVs), improved organic conducting and semiconducting materials are needed. The progress in two fields is reviewed in this article.

In the field of organic printable electronics, such as OLEDs and organic photovoltaics (OPVs), improved organic conducting and semiconducting materials are needed. The progress in two fields is reviewed in this article.

In the field of organic printable electronics, such as OLEDs and organic photovoltaics (OPVs), improved organic conducting and semiconducting materials are needed. The progress in two fields is reviewed in this article.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service