179523
Dimethyl suberimidate dihydrochloride
98%
Synonym(s):
DMS, Dimethyl ester octanediimidic acid dihydrochloride
About This Item
Recommended Products
Assay
98%
form
powder
mp
213-214 °C (lit.)
functional group
amine
SMILES string
Cl.COC(=N)CCCCCCC(=N)OC
InChI
1S/C10H20N2O2.2ClH/c1-13-9(11)7-5-3-4-6-8-10(12)14-2;;/h11-12H,3-8H2,1-2H3;2*1H
InChI key
ILKCDNKCNSNFMP-UHFFFAOYSA-N
Gene Information
rat ... Maoa(29253) , Maob(25750)
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Chemical modification of tyrosinase for coupling to produce biocatalytic enzymatic flow-through detectors
- Cross-linker for synthesis of collagen-calcium phosphate composites
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service