Skip to Content
Merck
All Photos(2)

Key Documents

D2388

Sigma-Aldrich

Dimethyl 3,3′-dithiopropionimidate dihydrochloride

powder

Synonym(s):

DTBP, Dimethyl 3,3′-dithio-bis(propionimidate) dihydrochloride, Wang/Richards’ reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2O2S2 · 2HCl
CAS Number:
Molecular Weight:
309.28
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reagent type: cross-linking reagent

solubility

H2O: 50 mg/mL

storage temp.

2-8°C

SMILES string

Cl.Cl.COC(=N)CCSSCCC(=N)OC

InChI

1S/C8H16N2O2S2.2ClH/c1-11-7(9)3-5-13-14-6-4-8(10)12-2;;/h9-10H,3-6H2,1-2H3;2*1H

InChI key

CQBCVFHLZAVNPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Synthesis of glutathione-sensitive cross-linked polyethylenimine gene vector
  • Preparation of cyclodextrin-containing polymers designed for gene delivery
  • Crosslinking of chick oviduct progesterone-receptor subunits

Caution

The reagant is readily hydrolyzed at neutral pH. Avoid use of reducing agents during coupling reactions.

Other Notes

Note that the amidine linkage preserves original primary amine positive charge. The disulfide linkage is cleavable by mild reduction. Incorporates an eight atom linker.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John G Lock et al.
Nature cell biology, 20(11), 1290-1302 (2018-10-27)
Adhesion to the extracellular matrix persists during mitosis in most cell types. However, while classical adhesion complexes, such as focal adhesions, do and must disassemble to enable mitotic rounding, the mechanisms of residual mitotic cell-extracellular matrix adhesion remain undefined. Here
Carine F Djuika et al.
Scientific reports, 7(1), 4410-4410 (2017-07-02)
Artemisinins are the current mainstay of malaria chemotherapy. Their exact mode of action is an ongoing matter of debate, and several factors have recently been reported to affect an early stage of artemisinin resistance of the most important human malaria
Lalit Yadav et al.
Organic & biomolecular chemistry, 18(30), 5927-5936 (2020-07-22)
A Bu4NI-catalyzed, DTBP-promoted, regioselective C(sp2)-C(sp3) cross dehydrogenative coupling (CDC) protocol for the direct C-3 benzylation of 2H-indazoles is reported. The metal-free protocol is operationally simple and proceeds mechanistically via the generation of stable benzylic free-radicals followed by regioselective addition at
Choong Eun Jin et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(10), 1800614-1800614 (2018-10-26)
Cell-free nucleic acids (cfNAs) are emerging diagnostic biomarkers for monitoring the treatment and recurrence of cancers. In particular, the biological role and clinical usefulness of cfNAs obtained from the plasma of patients with various cancers are popular and still intensely
Hyun Su Min et al.
Biomacromolecules, 19(6), 2320-2329 (2018-05-17)
Antibody fragment (Fab')-installed polyion complex (PIC) micelles were constructed to improve targetability of small interfering RNA (siRNA) delivery to pancreatic cancer cells. To this end, we synthesized a block copolymer of azide-functionalized poly(ethylene glycol) and poly(l-lysine) and prepared PIC micelles

Articles

Kanjiro Miyata (The University of Tokyo, Japan) provides insights on the rational design of polymeric materials for “smart” oligonucleotide delivery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service