Skip to Content
Merck
All Photos(1)

Key Documents

699713

Sigma-Aldrich

Isobutyltriethoxysilane

≥95%

Synonym(s):

Triethoxy(isobutyl)silane, Isobutyltriethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2Si(OC2H5)3
CAS Number:
Molecular Weight:
220.38
Beilstein:
1750049
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

liquid

refractive index

n20/D 1.4 (lit.)

bp

165 °C/400 mmHg (lit.)
190-191 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](CC(C)C)(OCC)OCC

InChI

1S/C10H24O3Si/c1-6-11-14(12-7-2,13-8-3)9-10(4)5/h10H,6-9H2,1-5H3

InChI key

ALVYUZIFSCKIFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

140.9 °F - closed cup

Flash Point(C)

60.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zijun Bie et al.
The Analyst, 144(9), 3128-3135 (2019-04-02)
Salbutamol (SAL) is one of the most widely abused feed additives in animal husbandry. Selective enrichment of SAL from complex biological samples is a key step for the analysis of SAL. Herein, we present SAL-imprinted magnetic nanoparticles (MNPs) as an
Weiman Zhao et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 234, 118258-118258 (2020-03-23)
Ginsenoside is a large family of triterpenoid saponins from Panax ginseng with various important biological functions. It is crucial to develop effective analytical approach for qualitative and quantitative analysis of ginsenosides. Herein, a dual boronate affinity nanoparticles-based plasmonic immunosandwich assay
Yueer Yan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(34), 12161-12170 (2015-07-16)
A clear and deep understanding of zeolite crystallization with the addition of organosilane is desirable for the reasonable design and preparation of hierarchical zeolites. Herein, the effects of different organosilanes on zeolite crystallization were systematically studied. It was found that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service