Merck
  • Safety assessment of Zanthoxylum alatum Roxb. essential oil, its antifungal, antiaflatoxin, antioxidant activity and efficacy as antimicrobial in preservation of Piper nigrum L. fruits.
Safety assessment of Zanthoxylum alatum Roxb. essential oil, its antifungal, antiaflatoxin, antioxidant activity and efficacy as antimicrobial in preservation of Piper nigrum L. fruits.
International journal of food microbiology (2011-12-06)
Bhanu Prakash, Priyanka Singh, Prashant Kumar Mishra, N K Dubey
ABSTRACT
The investigation deals with antifungal, antiaflatoxin and antioxidant efficacy of Zanthoxylum alatum Roxb. essential oil (EO), its two major constituents and their comparison with five commonly used organic acid preservatives. The chemical profile of EO, characterized through GC and GC-MS analysis, revealed linalool (56.10%) and methyl cinnamate (19.73%) as major components. The EO, linalool and methyl cinnamate completely inhibited the growth of a toxigenic strain of A. flavus (LHP-10) as well as aflatoxin B(1) secretion at different concentrations. Methyl cinnamate was found to be more efficacious than EO, linalool and five organic acid preservatives, showing antifungal and antiaflatoxigenic efficacy at a low concentration (0.6 μl/ml) and the nature of its toxicity was fungicidal. However, EO showed strong antioxidant activity with an IC(50) value at 5.6 μl/ml. Moreover, EO was found to have negligible mammalian toxicity as its LD(50) value, determined through oral administration on mice, was calculated to be 6124μl/kg body weight during safety profile assessment. During in vivo investigation on fruit systems, the Zanthoxylum EO, when tested as fumigant, provided 66.27% and 86.33% protection respectively at 1.25 μl/ml and 2.5 μl/ml against fungi infesting Piper nigrum L. fruits demonstrating its practical efficacy as a plant based antimicrobial for post harvest application.
Sigma-Aldrich
Methyl trans-cinnamate, 99%
Sigma-Aldrich
Methyl cinnamate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Methyl cinnamate, ≥99.0% (GC)
Sigma-Aldrich
Methyl trans-cinnamate, ≥98%, stabilized, FCC, FG