152625
5-Bromo-2-thiophenecarboxaldehyde
95%
Synonym(s):
5-Bromothenaldehyde
About This Item
Recommended Products
Assay
95%
refractive index
n20/D 1.637 (lit.)
bp
105-107 °C/11 mmHg (lit.)
density
1.607 g/mL at 25 °C (lit.)
functional group
aldehyde
storage temp.
2-8°C
SMILES string
[H]C(=O)c1ccc(Br)s1
InChI
1S/C5H3BrOS/c6-5-2-1-4(3-7)8-5/h1-3H
InChI key
GFBVUFQNHLUCPX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
210.2 °F - closed cup
Flash Point(C)
99.00 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service