151904
Ethanol-OD
≥99.5 atom % D
Synonym(s):
Ethan(ol-d), Ethyl alcohol-OD
About This Item
Recommended Products
isotopic purity
≥99.5 atom % D
Quality Level
Assay
≥99.00%
form
liquid
expl. lim.
3.5-15 % (lit.)
technique(s)
NMR: suitable
impurities
≤0.1000% water
water
refractive index
n20/D 1.359 (lit.)
bp
78 °C (lit.)
mp
-114 °C (lit.)
density
0.806 g/mL at 25 °C
mass shift
M+1
SMILES string
[2H]OCC
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3/i3D
InChI key
LFQSCWFLJHTTHZ-WFVSFCRTSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Recommended products
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
55.4 °F - closed cup
Flash Point(C)
13 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service