Skip to Content
Merck
All Photos(1)

Key Documents

151777

Sigma-Aldrich

Acetic acid-d

99 atom % D

Synonym(s):

mono-Deuteroacetic acid, Acetic (acid-d), Monodeuteroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOD
CAS Number:
Molecular Weight:
61.06
Beilstein:
1739249
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.07 (vs air)

Quality Level

vapor pressure

11.4 mmHg

isotopic purity

99 atom % D

Assay

99% (CP)

form

liquid

expl. lim.

4-19.9 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.50% water
water

bp

116-117 °C (lit.)

mp

15-16 °C (lit.)

density

1.059 g/cm3 at 25 °C

mass shift

M+1

SMILES string

[2H]OC(C)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i/hD

InChI key

QTBSBXVTEAMEQO-DYCDLGHISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetic acid-d (Monodeuteroacetic acid) is the monodeuterated form of acetic acid, in which the hydrogen atom of hydroxyl group has been replaced by deuterium (D). Infrared spectral studies of its vapors at 150°C in the range of 2-25μ have been reported.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Infrared Investigation of Acetic Acid and Acetic Acid-d Vapors and a Vibrational Assignment for the Monomeric Acids.
Wilmshurst JK.
J. Chem. Phys. , 25(6), 1171-1173 (1956)
Vennela Mullangi et al.
Biochemistry, 51(36), 7202-7208 (2012-08-21)
We report a method for expressing the solvent accessibility of histidine imidazole groups in proteins. The method is based on measuring the rate of the hydrogen exchange (HX) reaction of the imidazole C(ε1)-hydrogen. The rate profile of the HX reaction
Li-Min Peng et al.
PeerJ, 4, e1519-e1519 (2016-01-21)
According to myosatellite cell lines (MSCs) established in vitro from diploid and triploid flounder, we compared the characters of growth and differentiation of their MSCs. The results would be useful for learning the muscle development mechanism in teleosts. The skeletal
B R Miller et al.
Neuroscience, 153(1), 329-337 (2008-03-21)
The striatum, which processes cortical information for behavioral output, is a key target of Huntington's disease (HD), an autosomal dominant condition characterized by cognitive decline and progressive loss of motor control. Increasing evidence implicates deficient glutamate uptake caused by a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service